Methyl-5-bromo-2-chlorobenzoate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4570-5G CA-4570-25G CA-4570-1G
COBBCA-4570-5GEA 0 GBP
COBBCA-4570-5G COBBCA-4570-25G COBBCA-4570-1G
Methyl-5-bromo-2-chlorobenzoate
Methyl 5-bromo-2-chlorobenzoate
Formula: C₈H₆BrClO₂
MDL Number: MFCD00144770
CAS Number: 251085-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR