Methyl-2-quinolyl ether

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Methoxyquinoline

PY-1302-5G PY-1302-1G
COBBPY-1302-5GEA 0 GBP
COBBPY-1302-5G COBBPY-1302-1G
Methyl-2-quinolyl ether
Methyl 2-quinolyl ether
Formula: C₁₀H₉NO
MW: 159.19 g/mol
MDL Number: MFCD01070847
CAS Number: 6931-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR