7,8-Dichloroquinoline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2709-1G
HC-2709-5G
COBBHC-2709-1GEA
0
GBP
COBBHC-2709-1G
COBBHC-2709-5G
7,8-Dichloroquinoline
7,8-Dichloroquinoline
Formula:
C₉H₅Cl₂N |
MDL Number:
MFCD12026044 CAS Number: 703-49-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...