4,6-Dichloroquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6009-5G HC-6009-1G
COBBHC-6009-5GEA 0 GBP
COBBHC-6009-5G COBBHC-6009-1G
4,6-Dichloroquinoline
4,6-Dichloroquinoline
Formula: C₉H₅Cl₂N
MW: 198.05 g/mol
MDL Number: MFCD00156141
CAS Number: 4203-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR