5,8-Dibromoquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-8041-5G OT-8041-1G
COBBOT-8041-5GEA 0 GBP
COBBOT-8041-5G COBBOT-8041-1G
5,8-Dibromoquinoline
5,8-Dibromoquinoline
Formula: C₉H₅Br₂N
MW: 286.9507 g/mol
MDL Number: MFCD04966994
CAS Number: 81278-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR