2-tert-Butoxy-4-pyridineboronic acid pinacol ester
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-(tert-Butoxy)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
FM-2437-1G
FM-2437-250M
COBBFM-2437-1GEA
0
GBP
COBBFM-2437-1G
COBBFM-2437-250M
2-tert-Butoxy-4-pyridineboronic acid pinacol ester
2-tert-Butoxy-4-pyridineboronic acid pinacol ester
Formula:
C₁₅H₂₄BNO₃ MW: 277.1700 g/mol |
MDL Number:
MFCD11223286 CAS Number: 1256358-89-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...