1-Boc-indole-3-boronic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (1-(tert-Butoxycarbonyl)-1H-indol-3-yl)boronic acid

AS23379G1 AS23379G5
ACTIAS23379G1EA 0 GBP
ACTIAS23379G1 ACTIAS23379G5
1-Boc-indole-3-boronic acid
1-Boc-indole-3-boronic acid
Formula: C₁₃H₁₆BNO₄
MDL Number: MFCD06656257
CAS Number: 181365-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR