4-Bromo-1-ethoxy-2-nitrobenzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1268-5G AN-1268-1G
COBBAN-1268-5GEA 0 GBP
COBBAN-1268-5G COBBAN-1268-1G
4-Bromo-1-ethoxy-2-nitrobenzene
4-Bromo-1-ethoxy-2-nitrobenzene
Formula: C₈H₈BrNO₃
MDL Number: MFCD12756413
CAS Number: 383869-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR