2-Bromo-6-nitrobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS12103G1 AS12103G25 AS12103G5
ACTIAS12103G1EA 0 GBP
ACTIAS12103G1 ACTIAS12103G25 ACTIAS12103G5
2-Bromo-6-nitrobenzoic acid
2-Bromo-6-nitrobenzoic acid
Formula: C₇H₄BrNO₄
MDL Number: MFCD13688997
CAS Number: 38876-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR