2-Bromo-p-anisaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Bromo-4-methoxybenzaldehyde

AS32371G1 AS32371G5
ACTIAS32371G1EA 0 GBP
ACTIAS32371G1 ACTIAS32371G5
2-Bromo-p-anisaldehyde
2-Bromo-p-anisaldehyde
Formula: C₈H₇BrO₂
MW: 215.04 g/mol
MDL Number: MFCD09788636
CAS Number: 43192-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR