tert-Butyl-4-bromo-3-methoxybenzoate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4243-5G CA-4243-1G
COBBCA-4243-5GEA 0 GBP
COBBCA-4243-5G COBBCA-4243-1G
tert-Butyl-4-bromo-3-methoxybenzoate
tert-Butyl 4-bromo-3-methoxybenzoate
Formula: C₁₂H₁₅BrO₃
MW: 287.15 g/mol
MDL Number: MFCD08436063
CAS Number: 247186-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR