3-Bromopyridazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS32586G1 AS32586G5
ACTIAS32586G1EA 24 GBP
ACTIAS32586G1 ACTIAS32586G5
3-Bromopyridazine
3-Bromopyridazine
Formula: C₄H₃BrN₂
MW: 158.9840 g/mol
MDL Number: MFCD10688587
CAS Number: 88491-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR