3-Bromo-N-butyl-5-(trifluoromethyl)benzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6304-1G HC-6304-5G
COBBHC-6304-1GEA 0 GBP
COBBHC-6304-1G COBBHC-6304-5G
3-Bromo-N-butyl-5-(trifluoromethyl)benzenesulphonamide
3-Bromo-N-butyl-5-(trifluoromethyl)benzenesulphonamide
Formula: C₁₁H₁₃BrF₃NO₂S
MDL Number: MFCD09972137
CAS Number: 1020252-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR