3-Amino-N,N-dimethylbenzenesulphonamide
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Amino-N,N-Dimethyl-Benzenesulphonamide
AS22374G1
AS22374G25
AS22374G5
ACTIAS22374G1EA
129
GBP
ACTIAS22374G1
ACTIAS22374G25
ACTIAS22374G5
3-Amino-N,N-dimethylbenzenesulphonamide
3-Amino-N,N-dimethylbenzenesulphonamide
Formula:
C₈H₁₂N₂O₂S MW: 200.2600 g/mol |
MDL Number:
MFCD00602451 CAS Number: 6274-18-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...