tert-Butyl-2-chloro-5-fluorobenzoate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4357-5G CA-4357-1G
COBBCA-4357-5GEA 0 GBP
COBBCA-4357-5G COBBCA-4357-1G
tert-Butyl-2-chloro-5-fluorobenzoate
tert-Butyl 2-chloro-5-fluorobenzoate
Formula: C₁₁H₁₂ClFO₂
MDL Number: MFCD15143482
CAS Number: 1215206-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR