3,5-Dichloro-4-hydroxybenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Chloroxynil

AS24270G1 AS24270G25 AS24270G5
ACTIAS24270G1EA 88 GBP
ACTIAS24270G1 ACTIAS24270G25 ACTIAS24270G5
3,5-Dichloro-4-hydroxybenzonitrile
3,5-Dichloro-4-hydroxybenzonitrile
Formula: C₇H₃Cl₂NO
MW: 188.01 g/mol
MDL Number: MFCD00002177
CAS Number: 1891-95-8
EINECS: 217-572-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR