2-Chloro-6-hydroxybenzonitrile
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS12508G1
AS12508G5
ACTIAS12508G1EA
0
GBP
ACTIAS12508G1
ACTIAS12508G5
2-Chloro-6-hydroxybenzonitrile
2-Chloro-6-hydroxybenzonitrile
Formula:
C₇H₄ClNO MW: 153.5658 g/mol |
MDL Number:
MFCD01646165 CAS Number: 89999-90-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...