3-Bromo-N,N-dimethylbenzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0150-5G OT-0150-1G
COBBOT-0150-5GEA 0 GBP
COBBOT-0150-5G COBBOT-0150-1G
3-Bromo-N,N-dimethylbenzenesulphonamide
3-Bromo-N,N-dimethylbenzenesulphonamide
Formula: C₈H₁₀BrNO₂S
MW: 264.1395 g/mol
MDL Number: MFCD07363826
CAS Number: 153435-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR