3-Bromo-N-tert-butyl-N-methylbenzenesulphonamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0931-1G
OT-0931-250M
COBBOT-0931-1GEA
0
GBP
COBBOT-0931-1G
COBBOT-0931-250M
3-Bromo-N-tert-butyl-N-methylbenzenesulphonamide
3-Bromo-N-tert-butyl-N-methylbenzenesulphonamide
Formula:
C₁₁H₁₆BrNO₂S |
MDL Number:
MFCD12756462 CAS Number: 1187386-30-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...