2-Bromo-N,N-dimethylbenzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6134-5G HC-6134-25G HC-6134-1G
COBBHC-6134-5GEA 0 GBP
COBBHC-6134-5G COBBHC-6134-25G COBBHC-6134-1G
2-Bromo-N,N-dimethylbenzenesulphonamide
2-Bromo-N,N-dimethylbenzenesulphonamide
Formula: C₈H₁₀BrNO₂S
MW: 264.139 g/mol
MDL Number: MFCD07368229
CAS Number: 65000-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR