5-Bromo-2,4-difluorobenzenesulphonamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005530-2.5G 005530-1G
FLUO005530-2.5GEA 0 GBP
FLUO005530-2.5G FLUO005530-1G
5-Bromo-2,4-difluorobenzenesulphonamide
5-Bromo-2,4-difluorobenzenesulphonamide
Formula: C₆H₄BrF₂NO₂S
MDL Number: MFCD01940425
CAS Number: 287172-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR