1-tert-Butyl-6-fluorobenzimidazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HI-1513-1G HI-1513-5G
COBBHI-1513-1GEA 0 GBP
COBBHI-1513-1G COBBHI-1513-5G
1-tert-Butyl-6-fluorobenzimidazole
1-tert-Butyl-6-fluorobenzimidazole
Formula: C₁₁H₁₃FN₂
MDL Number: MFCD12546476
CAS Number: 1187385-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR