Methyl trifluoropyruvate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 3,3,3-trifluoropyruvate, Methyl 3,3,3-trifluoro-2-oxopropanoate

006851-5G 006851-25G 006851-100G
FLUO006851-5GEA 0 GBP
FLUO006851-5G FLUO006851-25G FLUO006851-100G
Methyl trifluoropyruvate
Methyl trifluoropyruvate
Formula: C₄H₃F₃O₃
MDL Number: MFCD00114936
CAS Number: 13089-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR