5-Bromo-8-methylquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6998-1G HC-6998-5G
COBBHC-6998-1GEA 0 GBP
COBBHC-6998-1G COBBHC-6998-5G
5-Bromo-8-methylquinoline
5-Bromo-8-methylquinoline
Formula: C₁₀H₈BrN
MDL Number: MFCD04966995
CAS Number: 74316-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR