5-Bromo-8-methylquinoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS25333G1 AS25333G5
ACTIAS25333G1EA 0 GBP
ACTIAS25333G1 ACTIAS25333G5
5-Bromo-8-methylquinoline
5-Bromo-8-methylquinoline
Formula: C₁₀H₈BrN
MDL Number: MFCD04966995
CAS Number: 74316-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR