2-Bromo-6-methylquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-1347-5G PY-1347-1G PY-1347-250M
COBBPY-1347-5GEA 0 GBP
COBBPY-1347-5G COBBPY-1347-1G COBBPY-1347-250M
2-Bromo-6-methylquinoline
2-Bromo-6-methylquinoline
Formula: C₁₀H₈BrN
MDL Number: MFCD01237982
CAS Number: 302939-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR