5-Fluoro-2-pyridineboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
5-Fluoropyridin-2-ylboronic acid
BB-5272-1G
BB-5272-250M
COBBBB-5272-1GEA
0
GBP
COBBBB-5272-1G
COBBBB-5272-250M
5-Fluoro-2-pyridineboronic acid
5-Fluoro-2-pyridineboronic acid
Formula:
C₅H₅BFNO₂ |
MDL Number:
MFCD08705712 CAS Number: 946002-10-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...