2,6-Difluoro-3-pyridineboronic acid hydrate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-8493-5G BB-8493-1G
COBBBB-8493-5GEA 0 GBP
COBBBB-8493-5G COBBBB-8493-1G
2,6-Difluoro-3-pyridineboronic acid hydrate
2,6-Difluoro-3-pyridineboronic acid hydrate
Formula: C₅H₄BF₂NO₂·H₂O
MDL Number: MFCD06657881
CAS Number: 1072952-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR