4-Amino-2-bromobenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24204G1 AS24204G25 AS24204G5
ACTIAS24204G1EA 0 GBP
ACTIAS24204G1 ACTIAS24204G25 ACTIAS24204G5
4-Amino-2-bromobenzonitrile
4-Amino-2-bromobenzonitrile
Formula: C₇H₅BrN₂
MDL Number: MFCD03695814
CAS Number: 53312-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR