2-Amino-3-bromobenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21298G1 AS21298G5
ACTIAS21298G1EA 0 GBP
ACTIAS21298G1 ACTIAS21298G5
2-Amino-3-bromobenzonitrile
2-Amino-3-bromobenzonitrile
Formula: C₇H₅BrN₂
MDL Number: MFCD05670539
CAS Number: 114344-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR