4-Amino-2-fluorobenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22861G1 AS22861G25 AS22861G5
ACTIAS22861G1EA 0 GBP
ACTIAS22861G1 ACTIAS22861G25 ACTIAS22861G5
4-Amino-2-fluorobenzonitrile
4-Amino-2-fluorobenzonitrile
Formula: C₇H₅FN₂
MDL Number: MFCD07368709
CAS Number: 53312-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR