7-Fluoro-1H-indazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21935G1 AS21935G25 AS21935G5
ACTIAS21935G1EA 0 GBP
ACTIAS21935G1 ACTIAS21935G25 ACTIAS21935G5
7-Fluoro-1H-indazole
7-Fluoro-1H-indazole
Formula: C₇H₅FN₂
MW: 136.1264 g/mol
MDL Number: MFCD09026976
CAS Number: 341-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR