3-Methyl-4-isoxazolecarboxylic acid ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
3-Methylisoxazole-4-carboxylic acid
L16307.03
L16307.MD
L16307.03EA
124
GBP
L16307.03
L16307.MD
3-Methyl-4-isoxazolecarboxylic acid ≥98%
3-Methyl-4-isoxazolecarboxylic acid
Purity: 98+%
CAS No.: 17153-20-7
Molecular Formula: C5H5NO3
Formula Weight: 127.10
Melting Point: 185-187°
Beilstein Reference: 114157
Formula:
C₅H₅NO₃ MW: 127.10 g/mol Melting Pt: 185…187 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318161 CAS Number: 17153-20-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...