2,5-Dichlorobenzoxazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21736G100 AS21736G25 AS21736G5
ACTIAS21736G100EA 0 GBP
ACTIAS21736G100 ACTIAS21736G25 ACTIAS21736G5
2,5-Dichlorobenzoxazole
2,5-Dichlorobenzoxazole
Formula: C₇H₃Cl₂NO
MDL Number: MFCD09264312
CAS Number: 3621-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR