3-Bromo-6-nitro-1H-indazole
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Bromo-6-nitroindazole
AS12300G25
AS12300G5
ACTIAS12300G25EA
0
GBP
ACTIAS12300G25
ACTIAS12300G5
3-Bromo-6-nitro-1H-indazole
3-Bromo-6-nitro-1H-indazole
Formula:
C₇H₄BrN₃O₂ MW: 242.0300 g/mol |
MDL Number:
MFCD06410881 CAS Number: 70315-68-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...