5-Bromo-3-nitro-o-toluic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22140G1 AS22140G5
ACTIAS22140G1EA 0 GBP
ACTIAS22140G1 ACTIAS22140G5
5-Bromo-3-nitro-o-toluic acid
5-Bromo-3-nitro-o-toluic acid
Formula: C₈H₆BrNO₄
MDL Number: MFCD07357316
CAS Number: 107650-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR