5-(3-Bromophenyl)-1H-tetrazole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OS-7678-5G
OS-7678-1G
COBBOS-7678-5GEA
0
GBP
COBBOS-7678-5G
COBBOS-7678-1G
5-(3-Bromophenyl)-1H-tetrazole
5-(3-Bromophenyl)-1H-tetrazole
Formula:
C₇H₅BrN₄ MW: 225.05 g/mol |
MDL Number:
MFCD01075675 CAS Number: 3440-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...