6-Bromo-1H-indazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2458-5G HC-2458-1G
COBBHC-2458-5GEA 0 GBP
COBBHC-2458-5G COBBHC-2458-1G
6-Bromo-1H-indazole
6-Bromo-1H-indazole
Formula: C₇H₅BrN₂
MDL Number: MFCD03265457
CAS Number: 79762-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR