4-Bromo-1H-indazole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2459-5G
HC-2459-1G
COBBHC-2459-5GEA
0
GBP
COBBHC-2459-5G
COBBHC-2459-1G
4-Bromo-1H-indazole
4-Bromo-1H-indazole
Formula:
C₇H₅BrN₂ |
MDL Number:
MFCD05664001 CAS Number: 186407-74-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...