Ethyl 4-iodobutyrate
Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Ethyl 4-iodobutanoate
ACTIAS25152G5EA
187
GBP
ACTIAS25152G5
ACTIAS25152G1
ACTIAS25152G25
Ethyl 4-iodobutyrate
Ethyl 4-iodobutyrate
Formula:
C₆H₁₁IO₂ MW: 242.0600 g/mol |
MDL Number:
MFCD00045325 CAS Number: 7425-53-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...