2-Amino-5-bromoisonicotinic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4139-5G
CA-4139-1G
COBBCA-4139-5GEA
0
GBP
COBBCA-4139-5G
COBBCA-4139-1G
2-Amino-5-bromoisonicotinic acid
2-Amino-5-bromoisonicotinic acid
Formula:
C₆H₅BrN₂O₂ MW: 217.0200 g/mol |
MDL Number:
MFCD09878357 CAS Number: 1000339-23-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...