4-(Chloromethyl)benzonitrile, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
726397-10G
726397-50G
SIAL726397-10GEA
88.8
GBP
SIAL726397-10G
SIAL726397-50G
4-(Chloromethyl)benzonitrile, Sigma-Aldrich®
4-(Chloromethyl)benzonitrile
Formula:
C₈H₆ClN MW: 151.60 g/mol |
MDL Number:
MFCD00019761 CAS Number: 874-86-2 EINECS: 212-869-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...