3-(3-Bromophenyl)propiononitrile

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B3199.5 B3199.25
XXXXB3199.5EA 0 GBP
XXXXB3199.5 XXXXB3199.25
3-(3-Bromophenyl)propiononitrile
3-(3-Bromophenyl)propiononitrile
Formula: C₉H₈BrN
MDL Number: MFCD01310794
CAS Number: 376646-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR