3-Fluoro-5-hydroxybenzoic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4337-5G
CA-4337-1G
CA-4337-25G
COBBCA-4337-5GEA
0
GBP
COBBCA-4337-5G
COBBCA-4337-1G
COBBCA-4337-25G
3-Fluoro-5-hydroxybenzoic acid
3-Fluoro-5-hydroxybenzoic acid
Formula:
C₇H₅FO₃ MW: 156.1112 g/mol |
MDL Number:
MFCD11846515 CAS Number: 860296-12-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...