2-Fluoro-3-hydroxybenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21934G1 AS21934G5
ACTIAS21934G1EA 0 GBP
ACTIAS21934G1 ACTIAS21934G5
2-Fluoro-3-hydroxybenzoic acid
2-Fluoro-3-hydroxybenzoic acid
Formula: C₇H₅FO₃
MDL Number: MFCD01862066
CAS Number: 91658-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR