Methyl-3-cyanobenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22043G100 AS22043G25
ACTIAS22043G100EA 0 GBP
ACTIAS22043G100 ACTIAS22043G25
Methyl-3-cyanobenzoate
Methyl 3-cyanobenzoate
Formula: C₉H₇NO₂
MDL Number: MFCD00220156
CAS Number: 13531-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR