4,5,6,7-Tetrahydro-1H-indazole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-3092-5G
HC-3092-1G
COBBHC-3092-5GEA
0
GBP
COBBHC-3092-5G
COBBHC-3092-1G
4,5,6,7-Tetrahydro-1H-indazole
Formula:
C₇H₁₀N₂ |
MDL Number:
MFCD00014565 CAS Number: 2305-79-5 EINECS: 218-977-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...