3-Amino-5-methylpyridazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23562G1 AS23562G5
ACTIAS23562G1EA 263 GBP
ACTIAS23562G1 ACTIAS23562G5
3-Amino-5-methylpyridazine
3-Amino-5-methylpyridazine
Formula: C₅H₇N₃
MW: 109.1292 g/mol
MDL Number: MFCD10698051
CAS Number: 144294-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR