3-Amino-4-methylpyridazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24092G1 AS24092G5
ACTIAS24092G1EA 431 GBP
ACTIAS24092G1 ACTIAS24092G5
3-Amino-4-methylpyridazine
3-Amino-4-methylpyridazine
Formula: C₅H₇N₃
MW: 109.1300 g/mol
MDL Number: MFCD08460975
CAS Number: 90568-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR