4-Ethoxy-3-(trifluoromethyl)aniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1324-5G AN-1324-25G AN-1324-1G
COBBAN-1324-5GEA 0 GBP
COBBAN-1324-5G COBBAN-1324-25G COBBAN-1324-1G
4-Ethoxy-3-(trifluoromethyl)aniline
4-Ethoxy-3-(trifluoromethyl)aniline
Formula: C₉H₁₀F₃NO
MW: 205.18 g/mol
MDL Number: MFCD08056137
CAS Number: 2713-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR